Learn More
US Plastic Corporation 6 Compartments Translucent K-Resin® Storage Box - 11" L x 6-3/4" W x 1-3/4" Hgt.
SDP
Catalog No. NC3521766
Encompass
Find items quickly in these clear boxes that are perfect for protecting and storing small items!K-Resin® Styrene Butadiene CopolymerScratch & abrasion resistantGreat impact resistanceHinged lid overlaps sides to protect contentsBoxes easily stackCompartment ID: (6) 6-1/4" L x 1-11/16" W x 1-9/16" Hgt.
Catalog No. NC3521766 Supplier US Plastic Corporation Supplier No. 65106
Find items quickly in these clear boxes that are perfect for protecting and storing small items!K-Resin® Styrene Butadiene CopolymerScratch & abrasion resistantGreat impact resistanceHinged lid overlaps sides to protect contentsBoxes easily stackCompartment ID: (6) 6-1/4" L x 1-11/16" W x 1-9/16" Hgt.