Learn More
Research Products International Corp Cardboard Storage Box with Lid, Standard 3 Inch, 5 1/4 x 5 1/4 x 2 7/8 Inches
SDP
Catalog No. 50212434
Encompass
RPI's cardboard storage box is made of heavy duty cardboard and is ideal for freezer and refrigerator storage. Boxes accept a cardboard cell partition to hold popular size micro-tubes (0.2 - 2.0 ml).
Helps maximize space
Boxes have a moisture resistant covering
Catalog No. 50-212-434 Supplier Research Products International Corp Supplier No. 181021
RPI's cardboard storage box is made of heavy duty cardboard and is ideal for freezer and refrigerator storage. Boxes accept a cardboard cell partition to hold popular size micro-tubes (0.2 - 2.0 ml).
Helps maximize space
Boxes have a moisture resistant covering