Learn More
Research Products International Corp STORAGE BOX, STD 3 INCH, ALUMINUM WITHOUT LID, 5 X 5 X 2 7/8 INCHES
SDP
Catalog No. 50212435
Encompass
Convenient storage system for 1.5ml-2.0ml micro-tubes and 1.2ml-2.0ml cryogenic vials. Stainless steel freezer racks available for both chest and upright freezers. Racks include stainless steel handles and accommodate all 2" and 3" high cardboard and metal storage boxes. Separate components allow system design compatibility with space requirements.
Catalog No. 50-212-435 Supplier Research Products International Corp Supplier No. 181023
Convenient storage system for 1.5ml-2.0ml micro-tubes and 1.2ml-2.0ml cryogenic vials. Stainless steel freezer racks available for both chest and upright freezers. Racks include stainless steel handles and accommodate all 2" and 3" high cardboard and metal storage boxes. Separate components allow system design compatibility with space requirements.