Learn More
Ted Pella Inc Plastic Box/Holder for 14 of 12.7mm dia. pin mounts
SDP
Catalog No. NC9231352
Encompass
Storage Box and Holder for 14 of 12.7mm dia. or 8 of ø25mm pin mounts (ø3.2mm - 1/8" pin). Numbered 1 - 14. Used for FEI/Philips, ZEISS/LEO/Leica, Cambridge, CamScan, Tescan, Amray and ETEC SEM's. Inside box dimensions are: 4-5/8" x 2-7/8" x 1-1/4" (11.7 x 7.3 x 3.2cm).
Catalog No. NC9231352 Supplier Ted Pella Inc Supplier No. 16717
Storage Box and Holder for 14 of 12.7mm dia. or 8 of ø25mm pin mounts (ø3.2mm - 1/8" pin). Numbered 1 - 14. Used for FEI/Philips, ZEISS/LEO/Leica, Cambridge, CamScan, Tescan, Amray and ETEC SEM's. Inside box dimensions are: 4-5/8" x 2-7/8" x 1-1/4" (11.7 x 7.3 x 3.2cm).