Learn More
Microsolv Technology Corporation Storage Box for 20ml EPA Style Vials. Violet, chemical resistant polypropylene holds 16 vials. Complete with translucent plastic lid. 1 EA.
Catalog No. 502271352
Encompass_Preferred
Excellent for standing position on the lab bench or during transport and ideal for space saving storage in places such as refrigerators. Lid prevents condensation on the caps minimizing contamination during storage. Usable temperature range -80C to +100C. Box outer dimensions are 130mm wide by 130mm deep by 102mm high. Box and lid are Autoclavable.
This item is not returnable. View return policy
Catalog No. 50-227-1352 Supplier Microsolv Technology Corporation Supplier No. 95040SB1
Excellent for standing position on the lab bench or during transport and ideal for space saving storage in places such as refrigerators. Lid prevents condensation on the caps minimizing contamination during storage. Usable temperature range -80C to +100C. Box outer dimensions are 130mm wide by 130mm deep by 102mm high. Box and lid are Autoclavable.