Learn More
Microsolv Technology Corporation Storage Box for 2ml Autosampler Vials. Natural, chemical resistant polypropylene holds 81 vials. Complete with translucent plastic lid. 1 EA.
Catalog No. 502271355
Encompass_Preferred
Excellent for standing position on the lab bench or during transport and ideal for space saving storage in places such as refrigerators. Lid prevents condensation on the caps minimizing contamination during storage. Usable temperature range -80C to +100C. Alpha numeric indexing coded on bottom of wells and along the sides of the box for clear sample identification. Box outer dimensions are 130mm wide by 130mm deep by 45mm high. Box and lid are Autoclavable.
This item is not returnable. View return policy
Catalog No. 50-227-1355 Supplier Microsolv Technology Corporation Supplier No. 9502SB06
Excellent for standing position on the lab bench or during transport and ideal for space saving storage in places such as refrigerators. Lid prevents condensation on the caps minimizing contamination during storage. Usable temperature range -80C to +100C. Alpha numeric indexing coded on bottom of wells and along the sides of the box for clear sample identification. Box outer dimensions are 130mm wide by 130mm deep by 45mm high. Box and lid are Autoclavable.