Learn More
Microsolv Technology Corporation Storage Box for Headspace Vials. Blue, polypropylene holds 25 vials. Complete with lid. 1 EA.
Catalog No. 502271219
Encompass_Preferred
These stackable, Storage Boxes are often used for 20mm Headspace Vials for storage or transportation. The Alphanumeric Coding on the boxes makes it easier for Sample Identification. The "egg crate" separators make it easy to keep vials apart. Supplied with a translucent plastic Lid that keeps condensation from forming on the Septa and minimizes dust or dirt. They are Chemically Resistant and can be Autoclaved. Temperature Range: -80⁰ to 100⁰C
This item is not returnable. View return policy
Catalog No. 50-227-1219 Supplier Microsolv Technology Corporation Supplier No. 95025SB
These stackable, Storage Boxes are often used for 20mm Headspace Vials for storage or transportation. The Alphanumeric Coding on the boxes makes it easier for Sample Identification. The "egg crate" separators make it easy to keep vials apart. Supplied with a translucent plastic Lid that keeps condensation from forming on the Septa and minimizes dust or dirt. They are Chemically Resistant and can be Autoclaved. Temperature Range: -80⁰ to 100⁰C