Learn More
Electron Microscopy Sciences Gatan 3View Pin Stub Storage Box
SDP
Catalog No. 501929108
Encompass
The Gatan storage box has been developed to store 28 of the Gatan 3View pin stubs. Made from clear polystyrene, it has two hinges and a snap-lock. Pins are held by a custom high density closed cell NBR foam with holes. The high density closed cell foam doesn't crumble and is elastic enough to allow repetitive loading and retrieving of the pins. Outside dimensions: 120 x 84 x 36 mm.
Catalog No. 50-192-9108 Supplier Electron Microscopy Sciences Supplier No. 7595908
The Gatan storage box has been developed to store 28 of the Gatan 3View pin stubs. Made from clear polystyrene, it has two hinges and a snap-lock. Pins are held by a custom high density closed cell NBR foam with holes. The high density closed cell foam doesn't crumble and is elastic enough to allow repetitive loading and retrieving of the pins. Outside dimensions: 120 x 84 x 36 mm.