Learn More
Alkali Scientific Tube Storage Box: Hinged Lid, Black, 100 x 1.5mL, 5/pk
SDP
Catalog No. 502389564
Encompass_Preferred
Suitable for refrigerator and freezer storage as low as -80˚C, Tube Storage Box have translucent lids and numbered cavities. Autoclavable. Five per pack.Suitable for refrigerator and freezer storage as low as -80˚C, Tube Storage Box have translucent lids and numbered cavities. Autoclavable. Five per pack.Suitable for refrigerator and freezer storage as low as -80˚C, Tube Storage Box have translucent lids and numbered cavities. Autoclavable. Five per pack.Suitable for refrigerator and freezer storage as low as -80˚C, Tube Storage Box have translucent lids and numbered cavities. Autoclavable. Five per pack.
Catalog No. 50-238-9564 Supplier Alkali Scientific Supplier No. FS100BK
Suitable for refrigerator and freezer storage as low as -80˚C, Tube Storage Box have translucent lids and numbered cavities. Autoclavable. Five per pack.Suitable for refrigerator and freezer storage as low as -80˚C, Tube Storage Box have translucent lids and numbered cavities. Autoclavable. Five per pack.Suitable for refrigerator and freezer storage as low as -80˚C, Tube Storage Box have translucent lids and numbered cavities. Autoclavable. Five per pack.Suitable for refrigerator and freezer storage as low as -80˚C, Tube Storage Box have translucent lids and numbered cavities. Autoclavable. Five per pack.