Learn More
Chemglass Life Sciences Storage Box Only, Standard, White, 5-3/4" x 5-3/4" x 4-3/4"
SDP
Catalog No. NC1390120
Encompass_Preferred
Storage Box Only, Standard, White, 5-3/4" x 5-3/4" x 4-3/4". Durable storage boxes made of cardboard treated with a moisture-repellent to protect from the effects of liquid nitrogen and condensation from freezer equipment. Boxes can withstand temperatures as low as -196¿C. Small drain holes in the bottom corners allow drainage of excess moisture. Designed to be compatible with most standard upright rack systems. Available white or yellow and in a variety of sizes / configurations to best match your needs.
Catalog No. NC1390120 Supplier Chemglass Life Sciences Supplier No. CLS1198W13
Storage Box Only, Standard, White, 5-3/4" x 5-3/4" x 4-3/4". Durable storage boxes made of cardboard treated with a moisture-repellent to protect from the effects of liquid nitrogen and condensation from freezer equipment. Boxes can withstand temperatures as low as -196¿C. Small drain holes in the bottom corners allow drainage of excess moisture. Designed to be compatible with most standard upright rack systems. Available white or yellow and in a variety of sizes / configurations to best match your needs.