Learn More
Sigma Aldrich Fine Chemicals Biosciences Storage box polycarbonate
Catalog No. 502901496
Encompass
Uncompartmented for storage of various size items in any environment from -196 to 121C. Can be washed and autoclaved repeatedly.
Catalog No. 50-290-1496 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. S70314EA
Uncompartmented for storage of various size items in any environment from -196 to 121C. Can be washed and autoclaved repeatedly.