Learn More
Ted Pella Inc Slide Storage File for 100 slides, white
SDP
Catalog No. NC9636541
Encompass
The Handy Slide Box has capacity of 100 3 x 1" or 75mm x 25mm slides, numbered index in hinged lid. Soft cork layer in bottom part. Stackable. This large capacity microscope slide box is available in 6 colors. Black is recommended for light sensitive specimens. Made of polystyrene. Outer dimensions: 21 x 17 x 3cm (8.3 x 6.7 x 1.18")
Catalog No. NC9636541 Supplier Ted Pella Inc Supplier No. 21021
The Handy Slide Box has capacity of 100 3 x 1" or 75mm x 25mm slides, numbered index in hinged lid. Soft cork layer in bottom part. Stackable. This large capacity microscope slide box is available in 6 colors. Black is recommended for light sensitive specimens. Made of polystyrene. Outer dimensions: 21 x 17 x 3cm (8.3 x 6.7 x 1.18")