Learn More
Research Products International Corp Cardboard Storage Box with Lid, Holds 50ml and 15ml Centrifuge Tubes, 5 3/4 x 5 3/4 Inches
SDP
Catalog No. 50550274
Encompass
Cardboard boxes are economical and constructed from durable cardboard. Store samples with ease in a moisture resistant environment. Capacity of the boxes varies from 16 x 50ml tubes to 36 x 15ml tubes when using the optional cell dividers sold separately.
Boxes feature a telescoping lid and height of the lid can vary from 2 7/8 inches to 5 1/4 inches.
Catalog No. 50-550-274 Supplier Research Products International Corp Supplier No. 247570
Cardboard boxes are economical and constructed from durable cardboard. Store samples with ease in a moisture resistant environment. Capacity of the boxes varies from 16 x 50ml tubes to 36 x 15ml tubes when using the optional cell dividers sold separately.
Boxes feature a telescoping lid and height of the lid can vary from 2 7/8 inches to 5 1/4 inches.