Learn More
Air Science Usa Vented Chemical Storage Cabinet, Benchtop, 34" Wide, 115V 60Hz
SDP
Catalog No. NC1837397
Encompass_Preferred
Safestore Vented Chemical Storage Cabinets, Benchtop, (34S), 34" Wide. Safestore Ductless Vented Chemical Storage Cabinets are useful for storage of noxious or odorous chemicals. These cabinets minimize health and environmental risks in handling chemical vapors and residues, VOCs and general laboratory functions. The Multiplex filtration system protects the safety of personnel during use, maintenance and decontamination. Safestore Features & Benefits: Activated carbon main filter formulated for general purpose storage to contain putrid organic odors as well as fumes (i.e. alcohol, solvents, gasoline, etc.). Safestore Vented Chemical Storage Cabinets brochure Perforated polypropylene shelves. With HEPA filter, eliminates exposure to biological particulates; see Accessories. Upflow airflow geometry operates. Includes qty. 1 Asts-001 Carbon Filter. Other Carbon and/or HEPA filters sold separately
Catalog No. NC1837397 Supplier Air Science Usa Supplier No. 34S
Safestore Vented Chemical Storage Cabinets, Benchtop, (34S), 34" Wide. Safestore Ductless Vented Chemical Storage Cabinets are useful for storage of noxious or odorous chemicals. These cabinets minimize health and environmental risks in handling chemical vapors and residues, VOCs and general laboratory functions. The Multiplex filtration system protects the safety of personnel during use, maintenance and decontamination. Safestore Features & Benefits: Activated carbon main filter formulated for general purpose storage to contain putrid organic odors as well as fumes (i.e. alcohol, solvents, gasoline, etc.). Safestore Vented Chemical Storage Cabinets brochure Perforated polypropylene shelves. With HEPA filter, eliminates exposure to biological particulates; see Accessories. Upflow airflow geometry operates. Includes qty. 1 Asts-001 Carbon Filter. Other Carbon and/or HEPA filters sold separately