Learn More
Grainger Durham Mfg Shelf Cabinet, 16-Gauge Steel, 48 in x 24 in x 72 in, 4 Adjustable Shelves, 1 ea
Catalog No. 502788449
Encompass_Preferred
This heavy-duty industrial storage cabinet is constructed from all-welded 16-gauge steel, providing a durable solution for organizing equipment and materials in warehouses, garages, packing stations, or shipping departments. The unit features two solid, recessed-style doors that open at 180° for full access to stored contents and includes a 3-point locking system for security.
  • Features 4 adjustable shelves that adjust on 4 in. centers
  • Provides a total load capacity of 3,000 lb. with a per-shelf capacity of 750 lb.
  • Finished in a textured, gray powder coat for increased durability
  • Includes 4 keyed chrome handles
  • Arrives fully assembled and features pre-punched holes for anchoring to the floor
Catalog No. 50-278-8449 Supplier Grainger Supplier No. 16D682
This heavy-duty industrial storage cabinet is constructed from all-welded 16-gauge steel, providing a durable solution for organizing equipment and materials in warehouses, garages, packing stations, or shipping departments. The unit features two solid, recessed-style doors that open at 180° for full access to stored contents and includes a 3-point locking system for security.
  • Features 4 adjustable shelves that adjust on 4 in. centers
  • Provides a total load capacity of 3,000 lb. with a per-shelf capacity of 750 lb.
  • Finished in a textured, gray powder coat for increased durability
  • Includes 4 keyed chrome handles
  • Arrives fully assembled and features pre-punched holes for anchoring to the floor