Learn More
Grainger Durham Manufacturing Storage Cabinet, 12 Gauge Steel, 8 Shelves, 1 ea
Catalog No. 502789937
Encompass_Preferred
This heavy-duty storage cabinet is designed for demanding industrial environments, providing a secure and organized solution for tools and equipment. Constructed from 12-gauge steel with a textured powder coat finish, this unit is built for durability and long-term use. It features a spacious interior with 8 adjustable shelves, allowing for flexible storage configurations to accommodate various item sizes.
  • Constructed from 12-gauge steel with 7-gauge leaf hinges for maximum durability
  • Includes 8 adjustable shelves for customizable storage space
  • Equipped with a 3-point locking system and a pad-lockable cast iron handle
  • Features a forkliftable design with 6 in. base legs for easy transport and positioning
  • Fully assembled for immediate use
  • Includes pre-punched holes to accommodate lagging to the floor
Catalog No. 50-278-9937 Supplier Grainger Supplier No. 16D697
This heavy-duty storage cabinet is designed for demanding industrial environments, providing a secure and organized solution for tools and equipment. Constructed from 12-gauge steel with a textured powder coat finish, this unit is built for durability and long-term use. It features a spacious interior with 8 adjustable shelves, allowing for flexible storage configurations to accommodate various item sizes.
  • Constructed from 12-gauge steel with 7-gauge leaf hinges for maximum durability
  • Includes 8 adjustable shelves for customizable storage space
  • Equipped with a 3-point locking system and a pad-lockable cast iron handle
  • Features a forkliftable design with 6 in. base legs for easy transport and positioning
  • Fully assembled for immediate use
  • Includes pre-punched holes to accommodate lagging to the floor