Learn More
Grainger Storage Cabinet HDPE Platinum 72 In
Catalog No. 502764367
Encompass_Preferred
The West Indigo 16x015 Storage Cabinet is a 72 inch tall HDPE plastic storage cabinet with two doors and three adjustable shelves. It has a platinum gray finish and is made from heavy duty, chemical resistant HDPE plastic. The cabinet has a weight capacity of 800 lbs per shelf. It measures 72 x 36 x 18 inches (H x W x D). The cabinet is ideal for industrial, commercial, or residential storage uses. It can be used to store tools, supplies, equipment, archived documents, or other items. The cabinet's doors can be locked for added security. Assembly is required upon delivery.
Catalog No. 50-276-4367 Supplier Grainger Supplier No. 16X015
The West Indigo 16x015 Storage Cabinet is a 72 inch tall HDPE plastic storage cabinet with two doors and three adjustable shelves. It has a platinum gray finish and is made from heavy duty, chemical resistant HDPE plastic. The cabinet has a weight capacity of 800 lbs per shelf. It measures 72 x 36 x 18 inches (H x W x D). The cabinet is ideal for industrial, commercial, or residential storage uses. It can be used to store tools, supplies, equipment, archived documents, or other items. The cabinet's doors can be locked for added security. Assembly is required upon delivery.