Learn More
Grainger Storage Cabinet HDPE Platinum 66 In
Catalog No. 502764219
Encompass_Preferred
The Grainger storage cabinets are made of steel with a powder coat finish for durability. They feature adjustable shelves to maximize storage space, 3-point locking handles for security, and leveling feet for stability on uneven floors. The cabinets come in various sizes, including 24" wide models that can fit easily in tight spaces. They are available in different colors like gray, black, blue and beige to match any workspace aesthetic. With high weight capacities over 1200 lbs, these heavy-duty cabinets are ideal for safely organizing tools, equipment, parts and other industrial supplies.
Catalog No. 50-276-4219 Supplier Grainger Supplier No. 16X009
The Grainger storage cabinets are made of steel with a powder coat finish for durability. They feature adjustable shelves to maximize storage space, 3-point locking handles for security, and leveling feet for stability on uneven floors. The cabinets come in various sizes, including 24" wide models that can fit easily in tight spaces. They are available in different colors like gray, black, blue and beige to match any workspace aesthetic. With high weight capacities over 1200 lbs, these heavy-duty cabinets are ideal for safely organizing tools, equipment, parts and other industrial supplies.