Learn More
Electron Microscopy Sciences Bench Top Storage/Dispenser Bin
SDP
Catalog No. 5030597
Encompass_Preferred
These workstation storage dispenser bins are fabricated from white and clear acrylic. The hinged top cover assures easy filling while the hinged front cover provides a generous opening for easy removal. All items that are stored remain dust-free, yet transparent through the clear front window.
Footprint: 8.5" (L) x 5.5"(W)
Height: 16"
Catalog No. 50-305-97 Supplier Electron Microscopy Sciences Supplier No. 6436416
These workstation storage dispenser bins are fabricated from white and clear acrylic. The hinged top cover assures easy filling while the hinged front cover provides a generous opening for easy removal. All items that are stored remain dust-free, yet transparent through the clear front window.
Footprint: 8.5" (L) x 5.5"(W)
Height: 16"