Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Storage File Systems
SDP

Catalog No. 22126344 Shop All Fisherbrand Products
Change view
Click to view available options
Slide Capacity:
2000
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Slide Capacity
22-126-344 -
22-126-343 2000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-126-344
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For long-term storage of histology microscope slide and tissue cassette specimens.

Fisherbrand™ slide and tissue cassette storage boxes provide an economical yet sturdy and secure method for long-term storage of your microscope slide and tissue cassette block specimens.

Our easy to assemble boxes come 10 per case and include 40 drawers/drawer inserts for the slide storage option (4 drawers per file) and 80 drawers/drawer inserts for the tissue block storage option (8 drawers per file).

The coated surface helps to ensure moisture resistance for long-term protection. Each microscope slide storage box holds up to 2000 microscope slides while each tissue block storage box holds up to 500 tissue blocks/cassettes. The unique locking tabs help keep stacked boxes safely secured and stable.

Specifications

Color White
Device Holds 500 Tissue Block
For Use With (Application) Histology Speciemen Storage
Height (Metric) 100 mm
Includes One file box and eight small drawers
Material Cardboard
Width (Metric) 299 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.