Learn More
World Precision Instrument Micropipette Storage Jar-E210
SDP
Catalog No. 50821852
Encompass
Store up to 30 micropipettes, filled or unfilled, up to three inches in length. A gentle sliding action inserts or removes pipettes without damage to the delicate tips.
Catalog No. 50-821-852 Supplier World Precision Instrument Supplier No. E210
Store up to 30 micropipettes, filled or unfilled, up to three inches in length. A gentle sliding action inserts or removes pipettes without damage to the delicate tips.