Learn More
Thomas Scientific STORAGE PLATE 48W 7.5ML 30/CS
SDP
Catalog No. NC2127458
Encompass
48-Well (8 rows x 6 columns) Polypropylene Storage/Reaction Microplate, 7.5mL/rectangle Well, pyramid bottoms, 68mm height
This item is not returnable. View return policy
Catalog No. NC2127458 Supplier Thomas Scientific Supplier No. 1149Q15
48-Well (8 rows x 6 columns) Polypropylene Storage/Reaction Microplate, 7.5mL/rectangle Well, pyramid bottoms, 68mm height