Learn More
Waters Corp MassTrak Endocrine Steroid Quality Control Set
Catalog No. 502269318 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The MassTrak Endocrine Steroid Quality Control Set is intended to be used in a clinical biochemistry laboratory by trained professionals to detect systematic analytical deviations that may arise from reagent or analytical instrument variation when performing the quantitative determination of steroids (mineralocorticoids, androgens, glucocorticoids and progestogens) in human serum using liquid chromatography – tandem mass spectrometry (LC-MS/MS) systems.The Waters MassTrak Endocrine Steroid Quality Control Set [one box] contains 3 matrix-matched controls in human serum and labeling. Contents include:• Quality Control Low (QC Low)• Quality Control Medium (QC Medium)• Quality Control High (QC High)• Insert Card referencing the IFU (Instructions for Use) SDS (Safety Data Sheets) & COA (Certification of Analysis)
This item is not returnable. View return policy
Catalog No. 50-226-9318 Supplier Waters Corp Supplier No. 186009312IVD
The MassTrak Endocrine Steroid Quality Control Set is intended to be used in a clinical biochemistry laboratory by trained professionals to detect systematic analytical deviations that may arise from reagent or analytical instrument variation when performing the quantitative determination of steroids (mineralocorticoids, androgens, glucocorticoids and progestogens) in human serum using liquid chromatography – tandem mass spectrometry (LC-MS/MS) systems.The Waters MassTrak Endocrine Steroid Quality Control Set [one box] contains 3 matrix-matched controls in human serum and labeling. Contents include:• Quality Control Low (QC Low)• Quality Control Medium (QC Medium)• Quality Control High (QC High)• Insert Card referencing the IFU (Instructions for Use) SDS (Safety Data Sheets) & COA (Certification of Analysis)