Learn More
Agilent Technologies 96-Well Storage/Reaction Microplate, Polypropylene, 0.5 mL, Irradiated, 25/Pk.
Catalog No. NC2057794
Encompass
These 96-well storage and reaction microplates are manufactured from polypropylene, offering excellent chemical resistance for a wide range of laboratory applications. The plates feature a 0.5 mL well volume with a round bottom design, optimized for reliable sample handling and storage. This irradiated version is designed to meet rigorous standards in the lab.
  • 96-well configuration with round wells
  • 0.5 mL capacity per well
  • Constructed from durable polypropylene
  • Round, spherical bottom design
  • Irradiated for specialized use
  • 14.3 mm plate height
Catalog No. NC2057794 Supplier Agilent Technologies Supplier No. 204601100
These 96-well storage and reaction microplates are manufactured from polypropylene, offering excellent chemical resistance for a wide range of laboratory applications. The plates feature a 0.5 mL well volume with a round bottom design, optimized for reliable sample handling and storage. This irradiated version is designed to meet rigorous standards in the lab.
  • 96-well configuration with round wells
  • 0.5 mL capacity per well
  • Constructed from durable polypropylene
  • Round, spherical bottom design
  • Irradiated for specialized use
  • 14.3 mm plate height