Learn More
Agilent Technologies Storage and Reaction Microplate, 96-Well, Polypropylene, 0.53 mL, 25/Pk.
Catalog No. NC2057793
Encompass
This 96-well storage and reaction microplate is manufactured from high-quality polypropylene and features a round well design with a spherical bottom. It is optimized for versatile laboratory applications requiring reliable sample storage or reaction environments.
  • Constructed from durable polypropylene material
  • Features 96 round wells with a spherical bottom
  • Includes a 0.53 mL total well volume
  • Measures 14.35 mm in height
  • Supplied in a 25/pk configuration
Catalog No. NC2057793 Supplier Agilent Technologies Supplier No. 204600100
This 96-well storage and reaction microplate is manufactured from high-quality polypropylene and features a round well design with a spherical bottom. It is optimized for versatile laboratory applications requiring reliable sample storage or reaction environments.
  • Constructed from durable polypropylene material
  • Features 96 round wells with a spherical bottom
  • Includes a 0.53 mL total well volume
  • Measures 14.35 mm in height
  • Supplied in a 25/pk configuration