Learn More
Boca Scientific Inc BOCARACK855 Freezer Storage System for 15ml and 50ml Conical Tubes, Freezer Rack with Tube Storage Boxes, Capacity of 64 50ml Tubes & 144 15ml Tubes
Catalog No. NC0234140
Encompass
BOCARACK Freezer Storage Systems provide a complete solution for storing biological samples in low temperature freezers. These sturdy stainless-steel racks include sliding drawers containing tube storage boxes. They are compatible with refrigerators and -80°C freezers. The Bocarack 855 contains 2 drawers with two types of boxes. One drawer has 4 boxes with a 4 x 4 configuration and a capacity of 16 50ml conical tubes each. The second drawer has 4 boxes with a 6 x 6 configuration and a capacity of 36 15ml conical tubes each. Numeric markings on the boxes provide a unique ID for every sample. The transparent cover allows you to see the samples and markings without removing the lid. The rack measures 24" deep x 6" wide x 10" high.
Catalog No. NC0234140 Supplier Boca Scientific Inc Supplier No. BOCARACK855
BOCARACK Freezer Storage Systems provide a complete solution for storing biological samples in low temperature freezers. These sturdy stainless-steel racks include sliding drawers containing tube storage boxes. They are compatible with refrigerators and -80°C freezers. The Bocarack 855 contains 2 drawers with two types of boxes. One drawer has 4 boxes with a 4 x 4 configuration and a capacity of 16 50ml conical tubes each. The second drawer has 4 boxes with a 6 x 6 configuration and a capacity of 36 15ml conical tubes each. Numeric markings on the boxes provide a unique ID for every sample. The transparent cover allows you to see the samples and markings without removing the lid. The rack measures 24" deep x 6" wide x 10" high.