Learn More
Sigma Aldrich Fine Chemicals Biosciences Storage rack for 50 and 10
Catalog No. 502904813
Encompass
Create a fi le system for 50- and 100-place slide boxes (see above)-storage rack makes organization of samples easy.Clear acrylic construction allows the labels to be seen on the slide boxesPlace boxes in rows or columns by color to create an easy coding systemBins 15Dimensions 711 x 171 x 228 mm
Catalog No. 50-290-4813 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. HS159890B
Create a fi le system for 50- and 100-place slide boxes (see above)-storage rack makes organization of samples easy.Clear acrylic construction allows the labels to be seen on the slide boxesPlace boxes in rows or columns by color to create an easy coding systemBins 15Dimensions 711 x 171 x 228 mm