Learn More
Agilent Technologies Storage/Reaction Microplate, 96-Well, Polypropylene, 2 mL, 25/Pk.
Catalog No. NC3138460
Encompass
This 96-well storage and reaction microplate is manufactured from ultrahigh purity polypropylene in a DNase/RNase-free cleanroom environment. Designed for reliable performance in high-throughput applications, the plate features square wells with pyramid bottoms and meets all ANSI/SLAS microplate standards.
  • Manufactured from ultrahigh purity polypropylene
  • 96-well format with square wells and pyramid bottoms
  • Total plate volume of 192 mL with 2.38 mL well volume
  • Irradiated for contamination control
  • Compliant with ISO 9001:2015 and ANSI/SLAS standards
  • Dimensions of 127.76 mm x 85.47 mm x 44.04 mm
Catalog No. NC3138460 Supplier Agilent Technologies Supplier No. 201379100
This 96-well storage and reaction microplate is manufactured from ultrahigh purity polypropylene in a DNase/RNase-free cleanroom environment. Designed for reliable performance in high-throughput applications, the plate features square wells with pyramid bottoms and meets all ANSI/SLAS microplate standards.
  • Manufactured from ultrahigh purity polypropylene
  • 96-well format with square wells and pyramid bottoms
  • Total plate volume of 192 mL with 2.38 mL well volume
  • Irradiated for contamination control
  • Compliant with ISO 9001:2015 and ANSI/SLAS standards
  • Dimensions of 127.76 mm x 85.47 mm x 44.04 mm