Learn More
Agilent Technologies Storage and Reaction Microplate, 96-Well, Polypropylene, Round Well, Spherical Bottom, 25/Pk.
Catalog No. NC2055315
Encompass
These 96-well storage and reaction microplates are designed for the efficient storage of chemical and biological samples. Molded from medical-grade, chemical-resistant polypropylene, these low-binding plates minimize biomolecule loss during storage and assays.
  • 96-well configuration with round wells and spherical bottoms
  • Manufactured from medical-grade, chemical-resistant polymers
  • Low-binding characteristics to reduce biomolecule loss
  • DNase/RNase-free and manufactured in an ISO 9001:2015 certified facility
  • Suitable for high-throughput screening, molecular biology, and genomics
  • 94% DNA recovery and 95% protein recovery
  • Compatible with heat seals or adhesive films
Catalog No. NC2055315 Supplier Agilent Technologies Supplier No. 203426100
These 96-well storage and reaction microplates are designed for the efficient storage of chemical and biological samples. Molded from medical-grade, chemical-resistant polypropylene, these low-binding plates minimize biomolecule loss during storage and assays.
  • 96-well configuration with round wells and spherical bottoms
  • Manufactured from medical-grade, chemical-resistant polymers
  • Low-binding characteristics to reduce biomolecule loss
  • DNase/RNase-free and manufactured in an ISO 9001:2015 certified facility
  • Suitable for high-throughput screening, molecular biology, and genomics
  • 94% DNA recovery and 95% protein recovery
  • Compatible with heat seals or adhesive films