Learn More
Agilent Technologies Storage/Reaction Microplate, 96-Well, Polypropylene, Square Well, V Bottom, 25/Pk.
Catalog No. NC3872371
Encompass
These 96-well storage and reaction microplates are designed for a variety of laboratory applications, including high-throughput screening in drug discovery, molecular biology, and genomic workflows. Molded from ultrahigh purity, medical-grade, and chemical-resistant polypropylene, these plates offer reliable performance and low-temperature durability for freezer or refrigerator storage.
  • Manufactured in an ISO 9001:2015 certified, DNase/RNase-free facility
  • Square well configuration with V-shaped conical bottoms
  • Capacity of 1.1 mL per well
  • Autoclavable at 121°C and 15 psi for 20 minutes
  • Irradiated for enhanced purity
  • Compatible with heat seals, adhesive films, and cap mats
  • Complies with ANSI/SLAS standards
  • Suitable for use with major automation platforms and benchtop equipment
Catalog No. NC3872371 Supplier Agilent Technologies Supplier No. 204355100
These 96-well storage and reaction microplates are designed for a variety of laboratory applications, including high-throughput screening in drug discovery, molecular biology, and genomic workflows. Molded from ultrahigh purity, medical-grade, and chemical-resistant polypropylene, these plates offer reliable performance and low-temperature durability for freezer or refrigerator storage.
  • Manufactured in an ISO 9001:2015 certified, DNase/RNase-free facility
  • Square well configuration with V-shaped conical bottoms
  • Capacity of 1.1 mL per well
  • Autoclavable at 121°C and 15 psi for 20 minutes
  • Irradiated for enhanced purity
  • Compatible with heat seals, adhesive films, and cap mats
  • Complies with ANSI/SLAS standards
  • Suitable for use with major automation platforms and benchtop equipment