Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Storage server (84TB)

Catalog No. A85210302 Shop All Epredia Products
Change view
Click to view available options
Product Type:
Storage Server
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
A85210302 Storage Server
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A85210302 Supplier Epredia™ Supplier No. A85210302
Add to Cart
Add to Cart
Request a Quote

Storage server (84TB) for use with the Epredia E1000 Dx whole-slide digital scanner.

  • Accessory for digital slide scanner

Specifications

Product Type Storage Server
For Use With (Equipment) Epredia E1000 Dx Whole-slide Digital Scanner

Epredia E1000 Dx Digital Pathology Solution has been cleared by the US FDA for In Vitro Diagnostic Use (K241717)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.