Learn More
Electron Microscopy Sciences Sample Screw Cap Vial Series 2, 1 dram, 15 x 45 mm
SDP
Catalog No. 5028435
Encompass_Preferred
Compact space savers offered in a variety of sizes. Ideal for the storage of liquids, powders, small specimens, etc.... Each set includes a partitioned box, vials with caps, labels, reference chart, and an exterior card holder and card. Alpha-numerical indexing system allows quick identification and location of stored items. The Screw Cap Vial Series has threaded glass vials with plastic lined caps. Arranged 5 x 20. Tray: 13"(L) x 3.5"(W) x 2.
Volume: 1 dram
Size: 15 x 45 mm
Catalog No. 50-284-35 Supplier Electron Microscopy Sciences Supplier No. 64265
Compact space savers offered in a variety of sizes. Ideal for the storage of liquids, powders, small specimens, etc.... Each set includes a partitioned box, vials with caps, labels, reference chart, and an exterior card holder and card. Alpha-numerical indexing system allows quick identification and location of stored items. The Screw Cap Vial Series has threaded glass vials with plastic lined caps. Arranged 5 x 20. Tray: 13"(L) x 3.5"(W) x 2.
Volume: 1 dram
Size: 15 x 45 mm