Learn More
Electron Microscopy Sciences Sample Screw Cap Vial Series 2, 2 dram, 17 x 60 mm
SDP
Catalog No. 5028436
Encompass_Preferred
Compact space savers offered in a variety of sizes. Ideal for the storage of liquids, powders, small specimens, etc.... Each set includes a partitioned box, vials with caps, labels, reference chart, and an exterior card holder and card. Alpha-numerical indexing system allows quick identification and location of stored items. The Screw Cap Vial Series has threaded glass vials with plastic lined caps. Arranged 5 x 20, tray: 11 1/4"(L) x 4 1/4"(W).
Volume: 2 dram
Size: 17 x 60 mm
Catalog No. 50-284-36 Supplier Electron Microscopy Sciences Supplier No. 64266
Compact space savers offered in a variety of sizes. Ideal for the storage of liquids, powders, small specimens, etc.... Each set includes a partitioned box, vials with caps, labels, reference chart, and an exterior card holder and card. Alpha-numerical indexing system allows quick identification and location of stored items. The Screw Cap Vial Series has threaded glass vials with plastic lined caps. Arranged 5 x 20, tray: 11 1/4"(L) x 4 1/4"(W).
Volume: 2 dram
Size: 17 x 60 mm