Learn More
Electron Microscopy Sciences Sample Storage Set Snap Cap Shell Vial Series, 4 dram, 25 x 52 mm
SDP
Catalog No. 5028438
Encompass_Preferred
Poly-plug, Knurled snap-caps are friction fitted with glass shell vial, straight sided, and a flat bottom. Each set includes a partitioned box, vials with caps, labels, reference chart, and an exterior card holder and card. Alpha-numerical indexing system allows quick identification and location of stored items. Tray size:10.5"(L) x 5.5"(W) x 2.5"(H).
Volume: 4 dram
Size: 25 x 52 mm
Catalog No. 50-284-38 Supplier Electron Microscopy Sciences Supplier No. 64277
Poly-plug, Knurled snap-caps are friction fitted with glass shell vial, straight sided, and a flat bottom. Each set includes a partitioned box, vials with caps, labels, reference chart, and an exterior card holder and card. Alpha-numerical indexing system allows quick identification and location of stored items. Tray size:10.5"(L) x 5.5"(W) x 2.5"(H).
Volume: 4 dram
Size: 25 x 52 mm