Learn More
Electron Microscopy Sciences Sample Storage Set Snap Cap Shell Vial Series 7, 7 dram, 29 x 65 mm
SDP
Catalog No. 5028439
Encompass_Preferred
Poly-plug, Knurled snap-caps are friction fitted with glass shell vial, straight sided, and a flat bottom. Each set includes a partitioned box, vials with caps, labels, reference chart, and an exterior card holder and card. Alpha-numerical indexing system allows quick identification and location of stored items. Tray size:13"(L) x 6 3/4"(L) x 3"(H).
Volume: 7 dram
Size: 29 x 65 mm
Catalog No. 50-284-39 Supplier Electron Microscopy Sciences Supplier No. 64278
Poly-plug, Knurled snap-caps are friction fitted with glass shell vial, straight sided, and a flat bottom. Each set includes a partitioned box, vials with caps, labels, reference chart, and an exterior card holder and card. Alpha-numerical indexing system allows quick identification and location of stored items. Tray size:13"(L) x 6 3/4"(L) x 3"(H).
Volume: 7 dram
Size: 29 x 65 mm