Learn More
Electron Microscopy Sciences Sample Storage Set Snap Cap Shell Vial Series, 3 dram, 12 x 114 mm
SDP
Catalog No. 5028437
Encompass_Preferred
Poly-plug, Knurled snap-caps are friction fitted with glass shell vial, straight sided, and a flat bottom. Each set includes a partitioned box, vials with caps, labels, reference chart, and an exterior card holder and card. Alpha-numerical indexing system allows quick identification and location of stored items. Tray size:13"(L)x 3.5"(W)x 5"(H).
Volume: 3 dram
Size: 12 x 114 mm
Catalog No. 50-284-37 Supplier Electron Microscopy Sciences Supplier No. 64276
Poly-plug, Knurled snap-caps are friction fitted with glass shell vial, straight sided, and a flat bottom. Each set includes a partitioned box, vials with caps, labels, reference chart, and an exterior card holder and card. Alpha-numerical indexing system allows quick identification and location of stored items. Tray size:13"(L)x 3.5"(W)x 5"(H).
Volume: 3 dram
Size: 12 x 114 mm