Learn More
Electron Microscopy Sciences Sample Storage Set Square Bottle Series 1, 1 oz
SDP
Catalog No. 5028441
Encompass_Preferred
A large size storage set. It comes with square clear glass bottles (1-ounce capacity)and black screw caps with plastic linings. Each set includes a partitioned box, vials with caps, labels, reference chart, and an exterior card holder and card. Alpha-numerical indexing system allows quick identification and location of stored items. Arrangement: 5 x 10, tray: 12 3/4"(L) x 6.5"(W) x 3 3/4"(H).
Volume: 1 oz
Catalog No. 50-284-41 Supplier Electron Microscopy Sciences Supplier No. 64281
A large size storage set. It comes with square clear glass bottles (1-ounce capacity)and black screw caps with plastic linings. Each set includes a partitioned box, vials with caps, labels, reference chart, and an exterior card holder and card. Alpha-numerical indexing system allows quick identification and location of stored items. Arrangement: 5 x 10, tray: 12 3/4"(L) x 6.5"(W) x 3 3/4"(H).
Volume: 1 oz