Learn More
Electron Microscopy Sciences Sample Storage Set Square Bottle Series 2, 2 oz
SDP
Catalog No. 5028442
Encompass_Preferred
A large size storage set. It comes with square clear glass bottles (1-ounce capacity)and black screw caps with plastic linings. Each set includes a partitioned box, vials with caps, labels, reference chart, and an exterior card holder and card. Alpha-numerical indexing system allows quick identification and location of stored items. Arrangement: 4x8, tray: 12"(L) x 6"(W) x 3.5"(H).
Volume: 2 oz
Catalog No. 50-284-42 Supplier Electron Microscopy Sciences Supplier No. 64282
A large size storage set. It comes with square clear glass bottles (1-ounce capacity)and black screw caps with plastic linings. Each set includes a partitioned box, vials with caps, labels, reference chart, and an exterior card holder and card. Alpha-numerical indexing system allows quick identification and location of stored items. Arrangement: 4x8, tray: 12"(L) x 6"(W) x 3.5"(H).
Volume: 2 oz