Learn More
Electron Microscopy Sciences Sample Storage Set Square Bottle Series 4, 4 oz
SDP
Catalog No. 5028440
Encompass_Preferred
A large size storage set. It comes with square clear glass bottles (1-ounce capacity)and black screw caps with plastic linings. Each set includes a partitioned box, vials with caps, labels, reference chart, and an exterior card holder and card. Alpha-numerical indexing system allows quick identification and location of stored items. Arrangement: 3x7, tray: 12.5"(L) x 5.5"(W) x 4.5"(H). Bottle
Volume: 4 oz
Catalog No. 50-284-40 Supplier Electron Microscopy Sciences Supplier No. 64283
A large size storage set. It comes with square clear glass bottles (1-ounce capacity)and black screw caps with plastic linings. Each set includes a partitioned box, vials with caps, labels, reference chart, and an exterior card holder and card. Alpha-numerical indexing system allows quick identification and location of stored items. Arrangement: 3x7, tray: 12.5"(L) x 5.5"(W) x 4.5"(H). Bottle
Volume: 4 oz