Learn More
Electron Microscopy Sciences Sample Storage Set Square Bottle Series, 8 oz
SDP
Catalog No. 5028444
Encompass_Preferred
A large size storage set. It comes with square clear glass bottles (1-ounce capacity)and black screw caps with plastic linings. Each set includes a partitioned box, vials with caps, labels, reference chart, and an exterior card holder and card. Alpha-numerical indexing system allows quick identification and location of stored items. Arrangement: 3x6, tray: 13"(L) x 6 3/4"(W) x 4 1/4"(H). Bottle
Volume: 8 oz
Catalog No. 50-284-44 Supplier Electron Microscopy Sciences Supplier No. 64284
A large size storage set. It comes with square clear glass bottles (1-ounce capacity)and black screw caps with plastic linings. Each set includes a partitioned box, vials with caps, labels, reference chart, and an exterior card holder and card. Alpha-numerical indexing system allows quick identification and location of stored items. Arrangement: 3x6, tray: 13"(L) x 6 3/4"(W) x 4 1/4"(H). Bottle
Volume: 8 oz