Learn More
Electron Microscopy Sciences Sample Storage Set Square Bottle Series, 1/4 oz
SDP
Catalog No. 5028443
Encompass_Preferred
A large size storage set. It comes with square clear glass bottles (1-ounce capacity)and black screw caps with plastic linings. Each set includes a partitioned box, vials with caps, labels, reference chart, and an exterior card holder and card. Alpha-numerical indexing system allows quick identification and location of stored items. Arrangement 5 x 10, tray:9-1⁄2" x 5" x 3-1⁄2".
Volume: 1/4 oz
Catalog No. 50-284-43 Supplier Electron Microscopy Sciences Supplier No. 64280
A large size storage set. It comes with square clear glass bottles (1-ounce capacity)and black screw caps with plastic linings. Each set includes a partitioned box, vials with caps, labels, reference chart, and an exterior card holder and card. Alpha-numerical indexing system allows quick identification and location of stored items. Arrangement 5 x 10, tray:9-1⁄2" x 5" x 3-1⁄2".
Volume: 1/4 oz