Learn More
HANNA INSTRUMENTS INC pH Electrode Storage Solution (500 mL)
SDP
Catalog No. NC1814909
Encompass
HI70300L is an electrode storage solution prepared with reagent grade chemicals that can be used to ensure optimum performance of your pH and ORP electrodes. Properly storing your pH electrode in a solution keeps the glass membrane well hydrated which maintains proper function and provides accurate readings.Special formulation to minimize bacterial growth on your electrode.Clearly marked expiration date and lot number for traceability.Designed for storing any pH or ORP electrode.
Catalog No. NC1814909 Supplier HANNA INSTRUMENTS INC Supplier No. HI70300L
HI70300L is an electrode storage solution prepared with reagent grade chemicals that can be used to ensure optimum performance of your pH and ORP electrodes. Properly storing your pH electrode in a solution keeps the glass membrane well hydrated which maintains proper function and provides accurate readings.Special formulation to minimize bacterial growth on your electrode.Clearly marked expiration date and lot number for traceability.Designed for storing any pH or ORP electrode.