Learn More
Cole-Parmer Oakton Electrode Storage Solution; 24 x 500 mL Bottles/Cs
Supplier:  Cole-Parmer 3565504
Encompass_Preferred
Remove grease and oil deposits from your electrodes with our cleaning solution. Cleaning solution contains pepsin. After cleaning, keep your electrodes in proper working condition with our storage solution.
Catalog No. 50-196-1366