Learn More
Grainger Rheem-Ruud Storage Tank, Vertical, Steel, 80 gal, 150 psi, 1 ea
Catalog No. 502757589
Encompass_Preferred
These commercial vertical storage tanks are designed to provide large quantities of hot water for commercial applications. Built for high-volume use, they can be installed individually or in parallel manifold configurations, offering an 80% draw capability without an appreciable decrease in temperature.
  • 80 gal. capacity with a durable steel tank construction
  • Proprietary steel formulation with two coats of high-temperature porcelain enamel for corrosion resistance
  • 2 in. rigid polyurethane foam insulation provided via a patented injection process to reduce operating costs
  • Includes two heavy-duty magnesium anode rods
  • Meets or exceeds current ASHRAE thermal efficiency and standby loss requirements
  • Maximum working pressure of 150 psi
  • Designed to pass through a 30 in. opening
Catalog No. 50-275-7589 Supplier Grainger Supplier No. 15A520
These commercial vertical storage tanks are designed to provide large quantities of hot water for commercial applications. Built for high-volume use, they can be installed individually or in parallel manifold configurations, offering an 80% draw capability without an appreciable decrease in temperature.
  • 80 gal. capacity with a durable steel tank construction
  • Proprietary steel formulation with two coats of high-temperature porcelain enamel for corrosion resistance
  • 2 in. rigid polyurethane foam insulation provided via a patented injection process to reduce operating costs
  • Includes two heavy-duty magnesium anode rods
  • Meets or exceeds current ASHRAE thermal efficiency and standby loss requirements
  • Maximum working pressure of 150 psi
  • Designed to pass through a 30 in. opening