Learn More
Cole-Parmer Akro-Mils 35-300 BLUE HDPE Storage Tote Bo x (without Lid), 3.7 cu ft, 26.63" x 18.25" x 14.88"
Catalog No. NC1185019 Shop All Cole-Parmer Products
Encompass_Preferred
Use these chemical resistant, high-density polyethylene tote boxes to carry or store chemicals or solvents. They are leak and spill resistant and its rounded interior edges allow for easy cleaning. To maximize storage space, boxes can be stacked on top of one another while used or nested inside one another when empty. Lids are sold separately.
Catalog No. NC1185019 Supplier Cole-Parmer Supplier No. 0681090
Use these chemical resistant, high-density polyethylene tote boxes to carry or store chemicals or solvents. They are leak and spill resistant and its rounded interior edges allow for easy cleaning. To maximize storage space, boxes can be stacked on top of one another while used or nested inside one another when empty. Lids are sold separately.