Learn More
Salimetrics LLC STORAGE TUBES 50/PK
Catalog No. NC0800150
Encompass
Swab Storage Tubes: 17 mm x 100 mm, 50/pack, ​Can be frozen (to -80° C), able to write on cap. Large outer tube, with a small insert and snap cap used for storing saliva samples collected via SalivaBio swab(s). The saturated SalivaBio Oral Swab (SOS) or SalivaBio Children's Swab (SCS)/SalivaBio Infant's Swab (SIS) is placed in the small insert and entire tube is frozen. Upon thawing and centrifuging the saliva is contained within the large outer tube.
Catalog No. NC0800150 Supplier Salimetrics LLC Supplier No. 5001.05
Swab Storage Tubes: 17 mm x 100 mm, 50/pack, ​Can be frozen (to -80° C), able to write on cap. Large outer tube, with a small insert and snap cap used for storing saliva samples collected via SalivaBio swab(s). The saturated SalivaBio Oral Swab (SOS) or SalivaBio Children's Swab (SCS)/SalivaBio Infant's Swab (SIS) is placed in the small insert and entire tube is frozen. Upon thawing and centrifuging the saliva is contained within the large outer tube.