Learn More
Chemglass Life Sciences 350mL Storage Vessel, Cajon, Airfree, Schlenk, 0-8mm Valve, 240mm OAH
SDP
Catalog No. AF009604
Encompass_Preferred
350mL Storage Vessel, Cajon, Airfree, Schlenk, 0-8mm Valve, 240mm OAH. Storage vessel sidearm has a 9mm OD tubulation for use with 3/8" Cajon connections. Chem-Vac™ Chem-Cap valve is used to control gas or liquid flow into the vessel.
Catalog No. AF009604 Supplier Chemglass Life Sciences Supplier No. AF009604
350mL Storage Vessel, Cajon, Airfree, Schlenk, 0-8mm Valve, 240mm OAH. Storage vessel sidearm has a 9mm OD tubulation for use with 3/8" Cajon connections. Chem-Vac™ Chem-Cap valve is used to control gas or liquid flow into the vessel.