Learn More
Newcomer Supply Out/In Cards for Slides, 1,000/box
SDP
Catalog No. NC1847051
Encompass
Out/In Cards mark when a slide has been removed for easy record keeping until it is returned to the file system. Constructed of a heavy pink cover stock, the slide Out/In Card is designed to help you better manage blocks that are removed from the storage box. Dimensions of card: 3 1/4" (H) x 1" (W). Designed to be used in the Economy (cardboard) storage drawers.
Catalog No. NC1847051 Supplier Newcomer Supply Supplier No. 6876PK
Out/In Cards mark when a slide has been removed for easy record keeping until it is returned to the file system. Constructed of a heavy pink cover stock, the slide Out/In Card is designed to help you better manage blocks that are removed from the storage box. Dimensions of card: 3 1/4" (H) x 1" (W). Designed to be used in the Economy (cardboard) storage drawers.