Learn More
Revvity Health Sciences Inc StorPlate 96-well, round U-bottom, polypropylene, 450µL, case of 200
Catalog No. 509050623
Encompass
StorPlate 96-well, round U-bottom, polypropylene, 450µL, case of 200
Catalog No. 50-905-0623 Supplier Revvity Health Sciences Inc Supplier No. 6008199
StorPlate 96-well, round U-bottom, polypropylene, 450µL, case of 200