Learn More
Cole-Parmer Ibi Scientific BDRAA115S Belly Dancer Shaker, 110/115VAC
Catalog No. NC9872897 Shop All Cole-Parmer Products
Encompass
Combination of vertical and horizontal orbital motion allows rapid exchange of solvents when staining and destaining gels and ensures lower background blotting. The lightweight and extremely quiet shaker has strong suction feet for load stability. Trouble free construction contains no belts or pulleys, keeping it simple and durable. High quality shunt wound DC motor and control allow for a long life and dependable performance. Variable speed control, from 0 to 125 rpm, with no cogging at slow speeds. Collar adjustment varies platform angle from flat to 12 °pitch. Orbital throw is 1 inch. The open-sided platform accommodates oversize containers. What's included: a platform, removable nonskid mat, 6-ft 110/115VAC power cord (no cord included for 220/240VAC model).
Catalog No. NC9872897 Supplier Cole-Parmer Supplier No. 5165000
Combination of vertical and horizontal orbital motion allows rapid exchange of solvents when staining and destaining gels and ensures lower background blotting. The lightweight and extremely quiet shaker has strong suction feet for load stability. Trouble free construction contains no belts or pulleys, keeping it simple and durable. High quality shunt wound DC motor and control allow for a long life and dependable performance. Variable speed control, from 0 to 125 rpm, with no cogging at slow speeds. Collar adjustment varies platform angle from flat to 12 °pitch. Orbital throw is 1 inch. The open-sided platform accommodates oversize containers. What's included: a platform, removable nonskid mat, 6-ft 110/115VAC power cord (no cord included for 220/240VAC model).